Learn More
Medchemexpress LLC Chitosan (MW 150000) | 9012-76-4 | 5 G
SDP
Catalog No. 5000411498
Encompass_Preferred
Chitosan (MW 150000) is a polycationic linear polysaccharide derived from chitin with a molecular weight of 150000. It is a versatile biomaterial due to its non-toxicity, low allergenicity, biocompatibility, and biodegradability.
  • Exhibits antitumor, antibacterial, antifungal, and antioxidant activities.
  • Reduces growth potential and induces pro-apoptotic effects in certain melanoma cells.
  • Low-molecular-weight chitosan can penetrate bacterial cell walls, inhibit DNA transcription and mRNA synthesis.
  • High-molecular-weight chitosan can bind to negatively charged components on the bacterial cell wall, altering cell permeability and blocking transport.
  • Can be used in water treatment, wound-healing materials, pharmaceutical excipients or drug carriers, obesity research, and as a scaffold for tissue engineering.
Catalog No. 50-004-11498 Supplier Medchemexpress LLC Supplier No. HYB2144A5G
Chitosan (MW 150000) is a polycationic linear polysaccharide derived from chitin with a molecular weight of 150000. It is a versatile biomaterial due to its non-toxicity, low allergenicity, biocompatibility, and biodegradability.
  • Exhibits antitumor, antibacterial, antifungal, and antioxidant activities.
  • Reduces growth potential and induces pro-apoptotic effects in certain melanoma cells.
  • Low-molecular-weight chitosan can penetrate bacterial cell walls, inhibit DNA transcription and mRNA synthesis.
  • High-molecular-weight chitosan can bind to negatively charged components on the bacterial cell wall, altering cell permeability and blocking transport.
  • Can be used in water treatment, wound-healing materials, pharmaceutical excipients or drug carriers, obesity research, and as a scaffold for tissue engineering.