Learn More
Medchemexpress LLC HY-B2144B 500mg Medchemexpress, Chitosan (MW 30000) CAS:9012-76-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2144B500MG
Encompass_Preferred
Medchemexpress, HY-B2144B 500mg Chitosan (MW 30000) CAS:9012-76-4 Chitosan (MW 30000) (Deacetylated chitin (MW 30000)) is a polycationic linear polysaccharide derived from chitin with the molecular weight of 30000. Chitosan is an versatile biomaterial because of its non-toxicity, low allergenicity, biocompatibility and biodegradability. Chitosan also has antitumor, antibacterial, antifungal, and antioxidant activities[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1253