Learn More
Medchemexpress LLC HY-112108 1g Medchemexpress, Chitosan oligosaccharide CAS:148411-57-8 Purity:>98%
SDP
Catalog No. 501969027
Encompass_Preferred
Medchemexpress, HY-112108 1g Chitosan oligosaccharide CAS:148411-57-8 Chitosan oligosaccharide (COS) is an oligomer of β-(1→4)-linked D-glucosamine. Chitosan oligosaccharide (COS) activates AMPK and inhibits inflammatory signaling pathways including NF-κB and MAPK pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9027 Supplier Medchemexpress LLC Supplier No. HY1121081G
Medchemexpress, HY-112108 1g Chitosan oligosaccharide CAS:148411-57-8 Chitosan oligosaccharide (COS) is an oligomer of β-(1→4)-linked D-glucosamine. Chitosan oligosaccharide (COS) activates AMPK and inhibits inflammatory signaling pathways including NF-κB and MAPK pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.