Learn More
Medchemexpress LLC Chitosan oligosaccharide (COS) | 148411-57-8 | 91.0% | 5 G
SDP
Catalog No. 5000436373
Encompass_Preferred
Chitosan oligosaccharide (COS) is an oligomer of β-(1→4)-linked D-glucosamine. It activates AMPK and inhibits inflammatory signaling pathways including NF-κB and MAPK pathways.
  • Activates AMPK.
  • Inhibits inflammatory signaling pathways including NF-κB and MAPK pathways.
  • Possesses anti-inflammation, anti-cancer, and anti-diabetes effects.
  • Interrupts cancer progression by modulating signaling proteins/pathways.
  • Induces death in various cancer cell types.
  • Suppresses production of proinflammatory cytokines.
  • Demonstrates anti-inflammatory effect on lymphocyte activation.
  • Inhibits UV-induced macroscopic appearance in mice skin, reducing signs of photoaging.
Catalog No. 50-004-36373 Supplier Medchemexpress LLC Supplier No. HY1121085G
Chitosan oligosaccharide (COS) is an oligomer of β-(1→4)-linked D-glucosamine. It activates AMPK and inhibits inflammatory signaling pathways including NF-κB and MAPK pathways.
  • Activates AMPK.
  • Inhibits inflammatory signaling pathways including NF-κB and MAPK pathways.
  • Possesses anti-inflammation, anti-cancer, and anti-diabetes effects.
  • Interrupts cancer progression by modulating signaling proteins/pathways.
  • Induces death in various cancer cell types.
  • Suppresses production of proinflammatory cytokines.
  • Demonstrates anti-inflammatory effect on lymphocyte activation.
  • Inhibits UV-induced macroscopic appearance in mice skin, reducing signs of photoaging.