Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chitosan oligosaccharide (COS) | 148411-57-8 | 91.0% | 500 MG
SDP

Catalog No. 5000436238
Encompass_Preferred

Chitosan oligosaccharide (COS) is an oligomer of β-(1→4)-linked D-glucosamine. It activates AMPK and inhibits inflammatory signaling pathways including NF-κB and MAPK pathways. It is for research use only and not sold to patients.

  • Oligomer of β-(1→4)-linked D-glucosamine
  • Activates AMPK
  • Inhibits inflammatory signaling pathways including NF-κB and MAPK pathways
  • For research use only
  • Appearance: solid
  • Color: light yellow to yellow
  • Structure classification: saccharides, polysaccharides
  • Initial source: endogenous metabolite
  • Solubility in DMSO: 50 mg/mL (needs ultrasonic)
  • Solubility in H2O: 50 mg/mL (needs ultrasonic)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-36238 Supplier Medchemexpress LLC Supplier No. HY112108500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chitosan oligosaccharide (COS) is an oligomer of β-(1→4)-linked D-glucosamine. It activates AMPK and inhibits inflammatory signaling pathways including NF-κB and MAPK pathways. It is for research use only and not sold to patients.

  • Oligomer of β-(1→4)-linked D-glucosamine
  • Activates AMPK
  • Inhibits inflammatory signaling pathways including NF-κB and MAPK pathways
  • For research use only
  • Appearance: solid
  • Color: light yellow to yellow
  • Structure classification: saccharides, polysaccharides
  • Initial source: endogenous metabolite
  • Solubility in DMSO: 50 mg/mL (needs ultrasonic)
  • Solubility in H2O: 50 mg/mL (needs ultrasonic)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.