Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chitotriose Trihydrochloride Hydrate 93.0+%, TCI America™
SDP

Catalog No. C264225MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C264225MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C264225MG Supplier TCI America Supplier No. C264225MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117436-78-9
Molecular Formula C18H38Cl3N3O13
Molecular Weight (g/mol) 610.86
InChI Key XTVOJWIUUCPOIF-ITRRTUPTSA-N
Synonym Chitosan Trimer
PubChem CID 133554226
IUPAC Name (3S,5S,6S)-5-amino-6-[(3S,5S,6S)-5-amino-6-[(3S,5S)-5-amino-4,6-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-2-(hydroxymethyl)oxane-3,4-diol;trihydrochloride
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)O)CO)CO)N)O)O)O.Cl.Cl.Cl

Specifications

Melting Point 255°C
Color White
Quantity 25 mg
Formula Weight 610.86
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Chitotriose Trihydrochloride Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.