Learn More
Enzo Life Sciences Chk 1 & 2 substrate (1mg)
SDP
Catalog No. 502011266
Encompass_Preferred
This peptide sequence, KKKVSRSGLYRSPSMPENLNRPR, corresponds to amino acids 206-225 of the phosphatase CDC25C, which is a phosphorylation target of Chk1 on Ser216. The addition of three N-terminal lysines allows the peptide to be used in phosphocellulose kinase assays. Both Chk1 and Chk2 will phosphorylate this peptide. Alternative name: CDC25C (206-225). Long Term Storage: -20°C.
Catalog No. 50-201-1266 Supplier Enzo Life Sciences Supplier No. BMLP1580001
This peptide sequence, KKKVSRSGLYRSPSMPENLNRPR, corresponds to amino acids 206-225 of the phosphatase CDC25C, which is a phosphorylation target of Chk1 on Ser216. The addition of three N-terminal lysines allows the peptide to be used in phosphocellulose kinase assays. Both Chk1 and Chk2 will phosphorylate this peptide. Alternative name: CDC25C (206-225). Long Term Storage: -20°C.