Learn More
TARGETMOL CHEMICALS INC CHK1-IN-3 5MG
SDP
Catalog No. 502581663
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CHK1-IN-3 is an inhibitor of checkpoint kinase 1 (CHK1; IC50 0.4 nM). purity: 100%
Catalog No. 50-258-1663 Supplier TARGETMOL CHEMICALS INC Supplier No. T107915MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CHK1-IN-3 is an inhibitor of checkpoint kinase 1 (CHK1; IC50 0.4 nM). purity: 100%