Learn More
Medchemexpress LLC CHK1-IN-4 hydrochloride | 00-00-0 | 480.75 g/mol | C18H19BrClN7O2 | 5 MG
SDP
Catalog No. 5000262901
Encompass_Preferred
CHK1-IN-4 hydrochloride is the hydrochloride salt of a small-molecule inhibitor of checkpoint kinase 1 (CHK1). It is used in preclinical biochemical and cell-based studies to inhibit CHK1 phosphorylation and evaluate antitumor activity. Supplied as a solid with defined solubility and storage recommendations for research use.
  • Potent checkpoint kinase 1 (CHK1) inhibitor active in tumor cells.
  • Hydrochloride salt form for improved solubility and handling.
  • Solid material; soluble in DMSO at 5 mg/mL with ultrasonic and warming.
  • Storage: in solvent -80°C for up to 6 months; sealed -20°C for up to 1 month.
  • Supplied in small research pack sizes suitable for preclinical studies.
Catalog No. 50-002-62901 Supplier Medchemexpress LLC Supplier No. HY128766A5MG
CHK1-IN-4 hydrochloride is the hydrochloride salt of a small-molecule inhibitor of checkpoint kinase 1 (CHK1). It is used in preclinical biochemical and cell-based studies to inhibit CHK1 phosphorylation and evaluate antitumor activity. Supplied as a solid with defined solubility and storage recommendations for research use.
  • Potent checkpoint kinase 1 (CHK1) inhibitor active in tumor cells.
  • Hydrochloride salt form for improved solubility and handling.
  • Solid material; soluble in DMSO at 5 mg/mL with ultrasonic and warming.
  • Storage: in solvent -80°C for up to 6 months; sealed -20°C for up to 1 month.
  • Supplied in small research pack sizes suitable for preclinical studies.