Learn More
Enzo Life Sciences Chk 1 monoclonal antibody (2G1D5) (100 µg)
SDP
Supplier:  Enzo Life Sciences ADI905770100
Encompass_Preferred
Clone: 2G1D2. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human Chk1. UniProt: O14757. Source: Purified from mouse ascites. Species Reactivity: Human, Mouse, Rat. Applications: WB. Recommended dilutions: Western Blot (1µg/ml, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-658-530