Learn More
Aviva Systems Biology Corp CHKA Antibody - middle region (ARP53583_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53583_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHKA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CHKA. Alternate names: CHK, CKI, CK, EKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61058