Learn More
ATCC CHLA-01-MED; Medulloblastoma; Human (Homo sapiens)
Supplier:  ATCC CRL3021
Encompass
CHLA-01-MED is a cell line exhibiting rounded morphology that was isolated from the brain of an 8-year-old male with medulloblastoma. Cells were isolated from the brain tumor resected at the time of diagnosis and were cultured in neurobasal medium after mechanical disruption. This cell line was deposited by the Children's Hospital of Los Angeles and can be used in neuroscience research.
Catalog No. 50-238-3282