Learn More
ATCC CHLA-06-ATRT; Brain Tumor; Human (Homo sapiens)
Catalog No. 502383288
Encompass
CHLA-06-ATRT is a cell line exhibiting rounded or attached morphology that was isolated from the cerebellum of a 4-month-old female with Atypical Teratoid Rhabdoid Tumor (ATRT). This cell line can be used for studying the metabolism of this tumor type and screening for therapeutic targets.
Catalog No. 50-238-3288 Supplier ATCC Supplier No. CRL3038
CHLA-06-ATRT is a cell line exhibiting rounded or attached morphology that was isolated from the cerebellum of a 4-month-old female with Atypical Teratoid Rhabdoid Tumor (ATRT). This cell line can be used for studying the metabolism of this tumor type and screening for therapeutic targets.