Learn More
Bioss Chlamydia LPS
Supplier: Bioss BSM72050M
Chlamydia LPS Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide