Learn More
ATCC Chlamydia trachomatis; Serovar Ba; Strain: Apache-2
Catalog No. 502382429
Encompass
Chlamydia trachomatis Serovar Ba strain Apache-2 is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated from the eye of a child from the Apache tribe in San Carlos, Arizona. This product has applications in sexually transmitted disease and trachoma research.
Catalog No. 50-238-2429 Supplier ATCC Supplier No. VR347
Chlamydia trachomatis Serovar Ba strain Apache-2 is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated from the eye of a child from the Apache tribe in San Carlos, Arizona. This product has applications in sexually transmitted disease and trachoma research.