Learn More
ATCC Chlamydia trachomatis; Serovar D; Strain: UW-3/Cx; Genomic DNA
Supplier:  ATCC VR885D
Encompass
Chlamydia DNA, in total DNA, was isolated from McCoy cells (ATCC CRL-1696) infected with Chlamydia trachomatis serovar D strain UW-3/Cx (ATCC VR-885). This product was prepared using methods known to inactivate infecting agent. It is suitable for use in PCR and other molecular biology applications. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-3958