Learn More
ATCC Chlamydia trachomatis; Serovar C; Strain TW-3
Catalog No. 502384104
Encompass
Chlamydia trachomatis strain TW-3 [Trachoma type C strain TW-3] is propagated in HEp-2 cells (ATCC CCL-23). This strain was isolated in Taiwan from a specimen taken from the human conjunctiva. Use this bacterium in your sexually transmitted disease research.
Catalog No. 50-238-4104 Supplier ATCC Supplier No. VR1477
Chlamydia trachomatis strain TW-3 [Trachoma type C strain TW-3] is propagated in HEp-2 cells (ATCC CCL-23). This strain was isolated in Taiwan from a specimen taken from the human conjunctiva. Use this bacterium in your sexually transmitted disease research.