Learn More
ATCC Chlamydia trachomatis; Serovar A; Strain: Har-13
Supplier:  ATCC VR571B
Encompass
Chlamydia trachomatis Trachoma type A strain HAR-13 [strain Har 13] is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated from the conjunctiva of a 4-year-old child with trachoma and was deposited by the Centers for Disease Control and Prevention.
Catalog No. 50-238-4835