Learn More
ATCC Chlamydia trachomatis; Serovar K; Strain: UW-31/Cx
Supplier:  ATCC VR887
Encompass
Chlamydia trachomatis trachoma type K strain UW-31/Cx is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated from the cervix of a human patient with cervicitis in Seattle, Washington, and has applications in sexually transmitted disease research.
Catalog No. 50-238-4679