Learn More
ATCC Chlamydia trachomatis; Serovar I; Strain: UW-12/Ur
Catalog No. 502382247
Encompass
Chlamydia trachomatis serovar I strain UW-12/Ur is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated from the urethra of a male with non-gonococcal urethritis and it has applications in sexually transmitted disease research.
Catalog No. 50-238-2247 Supplier ATCC Supplier No. VR880
Add to Cart
Add to Cart
Chlamydia trachomatis serovar I strain UW-12/Ur is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated from the urethra of a male with non-gonococcal urethritis and it has applications in sexually transmitted disease research.