Learn More
ATCC Chlamydia trachomatis; Serovar E; Strain: BOUR; Genomic DNA
Catalog No. 502382440
Encompass
Genomic DNA from Chlamydia trachomatis Serovar E strain BOUR was isolated from McCoy cells (ATCC CRL-1696) infected with Chlamydia trachomatis strain BOUR (ATCC VR-348B). This product was prepared using methods known to inactivate the infecting agent. The product can be used for PCR and other molecular applications. Source organism and host cells are also available through the ATCC Catalog.
Catalog No. 50-238-2440 Supplier ATCC Supplier No. VR348BD
Genomic DNA from Chlamydia trachomatis Serovar E strain BOUR was isolated from McCoy cells (ATCC CRL-1696) infected with Chlamydia trachomatis strain BOUR (ATCC VR-348B). This product was prepared using methods known to inactivate the infecting agent. The product can be used for PCR and other molecular applications. Source organism and host cells are also available through the ATCC Catalog.