Learn More
ATCC Chlamydia trachomatis; Serovar D; Strain: UW-3/Cx
Catalog No. 502382742
Encompass
Chlamydia trachomatis serovar D strain UW-3/Cx is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated from the cervix of an asymptomatic human patient. This product has applications in infectious disease and sexually transmitted disease research.
Catalog No. 50-238-2742 Supplier ATCC Supplier No. VR885
Chlamydia trachomatis serovar D strain UW-3/Cx is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated from the cervix of an asymptomatic human patient. This product has applications in infectious disease and sexually transmitted disease research.