Learn More
ATCC Chlamydia trachomatis; Serovar G; Strain: UW-57/Cx
Catalog No. 502382766
Encompass
Chlamydia trachomatis Serovar G strain UW-57/Cx is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated in 1971 from the epithelial tissue of a human cervix. This product has applications in infectious disease and sexually transmitted disease research.
Catalog No. 50-238-2766 Supplier ATCC Supplier No. VR878
Chlamydia trachomatis Serovar G strain UW-57/Cx is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated in 1971 from the epithelial tissue of a human cervix. This product has applications in infectious disease and sexually transmitted disease research.