Learn More
ATCC Chlamydia trachomatis; Serovar J; Strain: UW-36/Cx
Supplier:  ATCC VR886
Encompass
Chlamydia trachomatis trachoma type J strain UW-36/Cx is propagated in HeLa 229 cells (ATCC CCL-2.1). This strain was isolated in 1971 from the cervix of a human patient with cervicitis in Seattle, Washington, and was deposited by the Centers for Disease Control and Prevention.
Catalog No. 50-238-2773