Learn More
ATCC Chlamydia trachomatis; Serovar E; Strain: BOUR
Supplier:  ATCC VR348B
Encompass
Chlamydia trachomatis Serovar E strain BOUR is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated from an adult with probable clinical inclusion conjunctivitis and active trachoma and was deposited by the Centers for Disease Control and Prevention. This product has applications in sexually transmitted disease and trachoma research.
Catalog No. 50-238-3411