Learn More
ATCC Chlamydia trachomatis; Serovar H; Strain: UW-43/Cx
Supplier:  ATCC VR879
Encompass
Chlamydia trachomatis serovar H strain UW-43/Cx is propagated in McCoy [McCoy B] cells (ATCC CRL-1696). This strain was isolated in 1971 in Seattle, Washington, from the epithelial tissue of a human cervix. This bacterium has applications in sexually transmitted disease research.
Catalog No. 50-238-3776