Learn More
ATCC Chlamydia trachomatis; Strain: LGV I; DNA
Catalog No. 502383961
Encompass
Quantitative Genomic DNA from Chlamydia trachomatis LGV Serovar I strain 440 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-3961 Supplier ATCC Supplier No. VR901BD
Quantitative Genomic DNA from Chlamydia trachomatis LGV Serovar I strain 440 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.