Learn More
ATCC Chlamydia trachomatis; Strain: LGV III; DNA
Catalog No. 502384655
Encompass
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.
Catalog No. 50-238-4655 Supplier ATCC Supplier No. VR903D
ATCC Genuine Nucleics can be used for assay development, verification, validation, monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine bacterial load.