Learn More
ATCC Chlamydia trachomatis; Strain: UW-36/Cx; DNA
Catalog No. 502384608
Encompass
Chlamydia DNA, in total DNA, was isolated from HeLa-229 cells (ATCC CCL-2.1) infected with Chlamydia trachomatis strain UW-36/Cx (ATCC VR-886). This product was prepared using methods known to inactivate the infecting agent. It is suitable for use in PCR and other molecular biology applications. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-4608 Supplier ATCC Supplier No. VR886D
Chlamydia DNA, in total DNA, was isolated from HeLa-229 cells (ATCC CCL-2.1) infected with Chlamydia trachomatis strain UW-36/Cx (ATCC VR-886). This product was prepared using methods known to inactivate the infecting agent. It is suitable for use in PCR and other molecular biology applications. The source organism and host cells are also available through the ATCC catalog.