Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Chlamydia trachomatis Z055, LGVII434, Titered
SDP

Catalog No. 22158029 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-029 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-029 Supplier ZeptoMetrix Supplier No. 0801776
Add to Cart
Add to Cart

Used to determine limit of detection (LOD), in diagnostic assay development or crossreactivity studies

  • Each frozen aliquot contains 0.25mL of Chlamydia trachomatis that were propagated in monolayer of L929 cells
  • Titer is in Chlamydial inclusion forming units (IFU) and is measure of the infectivity of the material
  • Inclusions were quantified by direct immunofluorescence
  • Culture was frozen in sucrose-phosphate-glutamate buffer
  • Partially purified and may contain mouse cells
  • To maintain viability it is recommended to make any dilution of C. trachomatis with diluent buffer provided with material
  • Contain intact organisms and should be run in a manner identical to that used for clinical specimen

Specifications

Content And Storage -65°C or below
Format Titered
Product Type Titered Live Bacteria Control
Quantity 1 Each
Species Chlamydia trachomatis Z055, LGVII434
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.