Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Chlamydia trachomatis Z504; Jali20; serovar B, titered
SDP

Catalog No. 22158309 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-309 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-309 Supplier ZeptoMetrix Supplier No. 0804396
Add to Cart
Add to Cart

Live, titered microorganisms can be used to determine a limit of detection (LOD), in diagnostic assay development or cross reactivity studies.

  • Infectious, titered microorganisms can be utilized for analytical testing during assay development, including limit of detection (LOD) and cross-reactivity studies. When used as a control for nucleic acid tests, the same protocols as those used to extract and amplify clinical specimens should be employed.
  • Quality Control

Specifications

Content And Storage Titered material should be stored at -65°C or below
For Use With (Equipment) To avoid cross-contamination, use separate pipette tips for all reagents.
Includes Each frozen aliquot contains 0.25 mL of Chlamydia trachomatis that were propagated in a monolayer of McCoy cells. The titer is in Chlamydial inclusion forming units (IFU) and is a measure of the infectivity of the material.
No. per Pack 1 Each
Quantity 0.25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.