Learn More
ATCC Chlamydophila pneumoniae; Strain: CWL-029
Catalog No. 502381752
Encompass
Chlamydophila pneumoniae strain CWL-029 is propagated in HEp-2 cells (ATCC CCL-23). It was isolated from the throat swab of a pneumonia patient and deposited by the Centers for Disease Control and Prevention. Chlamydophila pneumoniae can be used in respiratory disease research.
Catalog No. 50-238-1752 Supplier ATCC Supplier No. VR1310
Chlamydophila pneumoniae strain CWL-029 is propagated in HEp-2 cells (ATCC CCL-23). It was isolated from the throat swab of a pneumonia patient and deposited by the Centers for Disease Control and Prevention. Chlamydophila pneumoniae can be used in respiratory disease research.