Learn More
ATCC Chlamydophila pneumoniae; Strain: 2023
Catalog No. 502382913
Encompass
Chlamydophila pneumoniae TWAR strain 2023 is propagated in HEp-2 cells (ATCC CCL-23). This bacterial strain was isolated from the nasopharynx of a human patient with pneumonia and can be used in infectious disease and respiratory disease research.
Catalog No. 50-238-2913 Supplier ATCC Supplier No. VR1356
Chlamydophila pneumoniae TWAR strain 2023 is propagated in HEp-2 cells (ATCC CCL-23). This bacterial strain was isolated from the nasopharynx of a human patient with pneumonia and can be used in infectious disease and respiratory disease research.