Learn More
ATCC Chlamydophila pneumoniae; Strain: AR-39
Catalog No. 502383002
Encompass
Chlamydophila pneumoniae strain AR-39 was isolated from the throat of an infected human with acute pharyngitis in Seattle, Washington. This bacterial strain is propagated in HEp-2 cells (ATCC CCL-23) and has applications in respiratory disease research.
Catalog No. 50-238-3002 Supplier ATCC Supplier No. 53592
Chlamydophila pneumoniae strain AR-39 was isolated from the throat of an infected human with acute pharyngitis in Seattle, Washington. This bacterial strain is propagated in HEp-2 cells (ATCC CCL-23) and has applications in respiratory disease research.