Learn More
ATCC Chlamydophila pneumoniae; Strain: CM-1
Supplier:  ATCC VR1360
Encompass
Chlamydophila pneumoniae strain CM-1 was isolated from the sputum of a pneumonia patient in Georgia, USA. This bacterial strain is propagated in HEp-2 cells (ATCC CCL-23) and has applications in respiratory disease research.
Catalog No. 50-238-3101