Learn More
ATCC Chlamydophila pneumoniae; Strain: TW-183
Supplier:  ATCC VR2282
Encompass
Chlamydophila pneumoniae strain TW-183 is propagated in HEp-2 cells (ATCC CCL-23). This strain was isolated in 1965 from the conjunctiva of a child in Taiwan. It has applications in respiratory and infectious disease research.
Catalog No. 50-238-4236
 Restricted Item