Learn More
ATCC CHL/IU (CHL-11) Lung; Chinese hamster (Cricetulus griseus)
Catalog No. 502382143
Encompass
CHL/IU [CHL-11] is a cell line exhibiting fibroblast-like morphology that was isolated from the lung of a female Chinese hamster. This cell line was deposited by M Ishidate and can be used in toxicology research.
Catalog No. 50-238-2143 Supplier ATCC Supplier No. CRL1935
CHL/IU [CHL-11] is a cell line exhibiting fibroblast-like morphology that was isolated from the lung of a female Chinese hamster. This cell line was deposited by M Ishidate and can be used in toxicology research.