Learn More
PALINTEST USA Kemio Chlorine Dioxide & Chlorite 6 months Refill Kit
Catalog No. NC2050112
Encompass
Kemio™ Chlorine Dioxide & Chlorite Refill Kit includes sensors and reagents for up to 6 months worth of chlorine dioxide and chlorite testing.Kit contents: 600 single-use CDX sensors, 2 x CR-1 reagent, 2 x CR-2 reagent, 2 x glycine reagent, 1 x pack of 10 stirring rods
Catalog No. NC2050112 Supplier PALINTEST USA Supplier No. KEM260CDX
Kemio™ Chlorine Dioxide & Chlorite Refill Kit includes sensors and reagents for up to 6 months worth of chlorine dioxide and chlorite testing.Kit contents: 600 single-use CDX sensors, 2 x CR-1 reagent, 2 x CR-2 reagent, 2 x glycine reagent, 1 x pack of 10 stirring rods