Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-Chloralose, 98+%, (max. 20% beta-anomer)

Catalog No. AC147791000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC147791000 100 g
AC147795000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC147791000 Supplier Thermo Scientific Chemicals Supplier No. 147791000

alpha-Chloralose, CAS #15879-93-3 is a true anesthetic or an immobilizing agent with sedative-hypnotic properties. | CAS: 15879-93-3 | C8H11Cl3O6 | 309.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • alpha-Chloralose is an anesthetic and an immobilizing agent
  • It involves autonomic reflexes

Applications

  • alpha-Chloralose is an anesthetic with sedative-hypnotic properties
  • It has been used in laboratory to anesthetize certain animals
  • It can be used to immobilize certain animals in the laboratory setting
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15879-93-3
Molecular Formula C8H11Cl3O6
Molecular Weight (g/mol) 309.52
MDL Number MFCD00005542
InChI Key OJYGBLRPYBAHRT-OPKHMCHVSA-N
Synonym a-chloralose, 1-2r,5r,6s,6ar-6-hydroxy-2-trichloromethyl-3a,5,6,6a-tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol, r-1-2r,3ar,5r,6s,6ar-6-hydroxy-2-trichloromethyl tetrahydrofuro 2,3-d 1,3 dioxol-5-yl ethane-1,2-diol
PubChem CID 7057995
IUPAC Name (1R)-1-[(2R,3aR,5R,6S,6aR)-6-hydroxy-2-(trichloromethyl)-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-5-yl]ethane-1,2-diol
SMILES OCC(O)[C@H]1OC2O[C@@H](O[C@@H]2[C@H]1O)C(Cl)(Cl)Cl

Specifications

Melting Point 176°C to 182°C
Color White to Yellow
CAS Min % 96.0
CAS Max % 100.0
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder
Assay Percent Range 97%
Merck Index 15, 2072
Formula Weight 309.52
Percent Purity ≥96.0%
Chemical Name or Material α-Chloralose, 97%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 240-016-7
RTECSNumber FM9450000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.