Learn More
Medchemexpress LLC HY-119417 100mg Medchemexpress, Chloramben CAS:133-90-4 Purity:>98%
SDP
Catalog No. 501969535
Encompass_Preferred
Medchemexpress, HY-119417 100mg Chloramben CAS:133-90-4 Chloramben (3-Amino-2,5-dichlorobenzoic acid) is a pre-emergence herbicide used to control the seedlings of annual grasses and broadleaf weeds. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9535 Supplier Medchemexpress LLC Supplier No. HY119417100MG
Medchemexpress, HY-119417 100mg Chloramben CAS:133-90-4 Chloramben (3-Amino-2,5-dichlorobenzoic acid) is a pre-emergence herbicide used to control the seedlings of annual grasses and broadleaf weeds. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.