Learn More
Crescent Chemical Co Inc CHLORAMBEN-METHYL SOLN 1ML
SDP
Catalog No. 5070543
Encompass_Preferred
Chloramben-methyl Single Component Calibration Standard in Methanol CAS 7286-84-2 Size - 1mL Catalog Number - 7239M.10
Catalog No. 50-705-43 Supplier Crescent Chemical Co Inc Supplier No. CT7239M.10
Chloramben-methyl Single Component Calibration Standard in Methanol CAS 7286-84-2 Size - 1mL Catalog Number - 7239M.10