Learn More
Medchemexpress LLC HY-13593 100mg , Chlorambucil CAS:305-03-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13593100MG
Encompass_Preferred
HY-13593 100mg Chlorambucil CAS: 305-03-3 Chlorambucil (CB-1348), an orally active antineoplastic agent, is a bifunctional alkylating agent belonging to the nitrogen mustard group. Chlorambucil can be used for the research of lymphocytic leukemia, ovarian and breast carcinomas, and Hodgkin’s disease[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1402