Learn More
TARGETMOL CHEMICALS INC Chlorambucil 100MG
SDP
Catalog No. 502880753
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Chlorambucil (CB-1348) is an orally-active antineoplastic aromatic nitrogen mustard. Chlorambucil alkylates and cross-links DNA during all phases of the cell cycle, resulting in disruption of DNA function, cell cycle arrest, and apoptosis. Purity 99.58%
Catalog No. 50-288-0753 Supplier TARGETMOL CHEMICALS INC Supplier No. T0975100MG
Also available in 1 mL, 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Chlorambucil (CB-1348) is an orally-active antineoplastic aromatic nitrogen mustard. Chlorambucil alkylates and cross-links DNA during all phases of the cell cycle, resulting in disruption of DNA function, cell cycle arrest, and apoptosis. Purity 99.58%