Learn More
Apexbio Technology LLC Chlorambucil 305-03-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063923
Encompass_Preferred
Chlorambucil (CAS 305-03-3) is an alkylating chemotherapeutic agent belonging to the nitrogen mustard class Its primary mechanism involves forming crosslinks within DNA strands both intra- and inter-strand crosslinks resulting in impaired DNA replication and transcription ultimately inducing cell death In vitro studies showed that chlorambucil potently inhibits human microvascular endothelial cells (IC 0 53 M) and exhibits cytotoxic activities against glioma cell lines such as SF767 (IC 114 M) and U87-MG (IC 96 M) Chlorambucil is widely used in research settings to investigate cell apoptosis DNA damage and lymphoid malignancies like chronic lymphocytic leukemia (CLL)
Catalog No. 50-000-63923 Supplier Apexbio Technology LLC Supplier No. B37165.1
Chlorambucil (CAS 305-03-3) is an alkylating chemotherapeutic agent belonging to the nitrogen mustard class Its primary mechanism involves forming crosslinks within DNA strands both intra- and inter-strand crosslinks resulting in impaired DNA replication and transcription ultimately inducing cell death In vitro studies showed that chlorambucil potently inhibits human microvascular endothelial cells (IC 0 53 M) and exhibits cytotoxic activities against glioma cell lines such as SF767 (IC 114 M) and U87-MG (IC 96 M) Chlorambucil is widely used in research settings to investigate cell apoptosis DNA damage and lymphoid malignancies like chronic lymphocytic leukemia (CLL)