Learn More
Cayman Chemical Chlorambucil, Purity: ≥98%, CAS: 305-03-3, 500 mg
SDP
Catalog No. 502994278
Encompass_Preferred
A DNA alkylating agent; cross-links DNA at 1 mg/ml and targets guanine-rich regions; reduces fetal brain size and induces microcephaly when administered to pregnant mice at 8 and 15 mg/kg
Catalog No. 50-299-4278 Supplier Cayman Chemical Supplier No. 23744500 MG
A DNA alkylating agent; cross-links DNA at 1 mg/ml and targets guanine-rich regions; reduces fetal brain size and induces microcephaly when administered to pregnant mice at 8 and 15 mg/kg