Learn More
Cayman Chemical Chlorambucil-d8, Purity: ≥99% deuterated forms (d1-d8), CAS: 2748247-26-7, 1 mg
SDP
Catalog No. 502934054
Encompass_Preferred
An internal standard for the quantification of chlorambucil by GC- or LC-MS
Catalog No. 50-293-4054 Supplier Cayman Chemical Supplier No. 267741 MG
An internal standard for the quantification of chlorambucil by GC- or LC-MS