Learn More
Cayman Chemical ChlorambucIl-d8 500ug
SDP
Catalog No. 502932965
Encompass_Preferred
An internal standard for the quantification of chlorambucil by GC- or LC-MS
Catalog No. 50-293-2965 Supplier Cayman Chemical Supplier No. 26774500 UG
An internal standard for the quantification of chlorambucil by GC- or LC-MS