Learn More
Medchemexpress LLC HY-B0959 1g , Chloramine-T CAS:127-65-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB09591G
Encompass_Preferred
HY-B0959 1g Chloramine-T CAS: 127-65-1 Chloramine-T is a titrimetric reagent, and an oxidizing agent. Chloramine-T is an oxidizing biocide. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1532