Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloramine-T Hydrate

Catalog No. 857319100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
100 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-731-9100G 1 g
85-731-95G 1 kg
85-731-91KG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 85-731-9100G Supplier Sigma Aldrich Supplier No. 857319100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chloramine-T Hydrate

Specifications

CAS 2018-61-3
Melting Point 171°C to 173°C
Molecular Formula C11H13NO3
Linear Formula C6H5CH2CH(NHCOCH3)CO2H
MDL Number MFCD00063158
Quantity 1 g
Synonym N-Chloro-p-toluenesulfonamide sodium salt hydrate
Molecular Weight (g/mol) 207.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.