Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloramine T sodium salt trihydrate, 100.29%, MP Biomedicals™

Catalog No. ICN15011591 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15011591 1 kg
ICN15011590 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15011591 Supplier MP Biomedicals, Inc Supplier No. 0215011591
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 127-65-1
Molecular Formula C7H7ClNNaO2S
Molecular Weight (g/mol) 227.638
InChI Key VDQQXEISLMTGAB-UHFFFAOYSA-N
Synonym chloramine-t, chloramine t, chloralone, chlorasan, chlorozone, acti-chlore, chloraseptine, chlorazene, chlorazone, chlorseptol
PubChem CID 3641960
ChEBI CHEBI:53767
IUPAC Name sodium;chloro-(4-methylphenyl)sulfonylazanide
SMILES CC1=CC=C(C=C1)S(=O)(=O)[N-]Cl.[Na+]

Specifications

Color White
Density Bulk density 540 to 680kg/m3
Assay Percent Range 1.0029
Quantity 1 kg
Formula Weight 281.7
Physical Form Fine Powder
Chemical Name or Material N-Chloro-p-toluenesulfonamide
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.